Vacuum filtering flasks, Büchner, with side arm socket, DURAN®
Supplier: DWK Life Sciences
211833603 211834402 211835407 211836309 211936804 211937303 211938608 211938805 211939107
519-4132EA 43.7 EUR
519-4132 519-4133 519-4134 519-4135 519-4231 519-4232 519-4233 519-4234 519-4235
Vacuum filtering flasks, Büchner, with side arm socket, DURAN®
Flasks Filter Flasks
DURAN®, borosilicate glass 3.3, clear.
  • Up to 2000 ml in Erlenmeyer shape, from 3000 ml in bottle shape
  • Ground socket to accommodate vacuum tubing of 15 to 18 mm ext.Ø (e.g. 6×5 or 8×5 mm)
  • Fulfils 'Equipment and product safety regulations'
For vacuum use. Filtration flasks with side arm socket have not only made work in preparation and analytical laboratories easier and simpler, but have also reduced accident risk.
DIN 12476 and ISO 6556 (only Erlenmeyer shape)
Certifications: ISO 6556 (Erlenmeyer shape) DIN 12475 (bottle shape).