Flasks, iodine without collar, with hollow glass stopper
Supplier: Lenz Laborglas GmbH & CO.KG
03026037 03026049 03026058 03026052 03026070
201-1261EA 17.2 EUR
201-1261 201-1262 201-1263 201-1038 201-1039
Flasks, iodine without collar, with hollow glass stopper
Flasks Iodine Flasks
DURAN®, borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Volume graduation to allow approximate reading of contents and labelling area.