Flasks, round bottom with three necks and Quickfit® ground joints
Supplier: SciLabware
FR250/3S/22A FR500/3S/22A FR100/2S/11A FR100/3S/11A FR250/4S/22A FR500/4S/22A FR250/2S/22A FR2L/3US/22A FR100/3S/22A FR25/1S/11A FR50/1S/11A FR1L/3S/22A FR50/2S/11A FR250/3S/11A FR250/4S/11A FR2L/3US/22A_U FR1L/4S/44A FR25/2S/11A FR50/2S/22A FR500/3S/11A FR1L/4S/22A FR2L/5S/22A FR3L/5S/22A
201-8506EA 78.2 EUR
201-8506 201-8539 201-8470 201-8472 201-8043 201-8047 201-2356 201-8513 201-2355 201-0610 201-0611 201-8487 201-0612 201-0712 BIBBFR250/4S/11A BIBBFR2L/3US/22A_U BIBBFR1L/4S/44A BIBBFR25/2S/11A BIBBFR50/2S/22A BIBBFR500/3S/11A BIBBFR1L/4S/22A BIBBFR2L/5S/22A BIBBFR3L/5S/22A
Flasks, round bottom with three necks and Quickfit® ground joints
Flasks Round Bottom Flasks
PYREX®, borosilicate glass, with inclined side necks, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
One centre neck and two side necks, both inclined at an angle of 20°. Matt white labelling area.
Certifications: ISO 383.