Flasks, round bottom, with two necks and standard ground joints
Supplier: Lenz Laborglas GmbH & CO.KG
03213137 03213237 03213249 03213258 03214137 03214149 03214158 03214170 03214173 03214449 03214458 03214470 03214473 03214479 03214237 03214270 03214258 03214249 03214273 03211122 03211128 03211137 03211149 03214437 03213270
201-1190EA 28.1 EUR
201-1190 201-0329 201-0330 201-0331 201-1404 201-1406 201-1407 201-1410 271-1412 271-1407 271-1409 271-1411 201-0333 201-0334 LENZ03214237 LENZ03214270 LENZ03214258 LENZ03214249 LENZ03214273 201-1401 201-1402 271-1403 271-1405 201-0332 201-1191
Flasks, round bottom, with two necks and standard ground joints
Flasks Round Bottom Flasks
DURAN®, borosilicate glass, with angled side neck, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Standard ground joints on the centre neck and the side neck, which is inclined at an angle of 20°. Matt white labelling area.