Volumetric flasks, class A, amber markings, BLAUBRAND® ETERNA
Supplier: Brand
36853 36854 36849 36847 36852 36838 36842 36843 36845 36848 36850 36851 36846 36841
612-5072EA 56.1 EUR
612-5072 612-2735 612-5068 612-5066 612-5071 612-5062 612-5063 612-5064 612-5065 612-5067 612-5069 612-5070 612-3652 612-3651
Volumetric flasks, class A, amber markings, BLAUBRAND® ETERNA
Flasks Volumetric Flasks
Borosilicate glass 3.3, with PP stopper.
  • Calibrated to contain (TC, In)
  • Mark and inscriptions in ETERNA amber stain
  • DE-M marking
*Wide neck.
Certifications: DIN EN ISO 1042
Ordering information: Supplied with a batch certificate. Volumetric flasks also available on request with an individual certificate, a USP individual certificate or a DAkkS calibration certificate.