Funnels with short stem
Supplier: GLASWARENFABRIK KARL HECHT
44000040 44000044 44000048 44000056 44000004 44000064 44000008 44000012 44000016 44000020 44000024 44000028 44000032 44000036
221-0126EA 6.3 EUR
221-0126 221-0127 221-0128 221-0129 221-0130 221-1146 221-0131 221-0132 221-0133 221-1135 221-0134 221-1137 221-0135 221-1139
Funnels with short stem
Funnels Filtration Funnels Filter Funnels
Borosilicate glass 3.3.
  • Good chemical and physical properties
  • Not suitable for applications involving high thermal stress
  • Quick and easy to clean
With smooth internal walls.
Certifications: DIN ISO 4798