Büchner funnels
Supplier: MORGAN ADVANCED MATERIALS HALD
L127C/0 L127C/00 L127C/0A L127C/1 L127C/2 L127C/2A L127C/3 L127C/4 L127C/4A L127C/5 L127C/6 L127C/6A L127C/8
511-2500EA 52.5 EUR
511-2500 511-2501 511-2503 511-2504 511-2505 511-2506 511-2507 511-2508 511-2509 511-2510 511-2511 511-2512 511-2513
Büchner funnels
Funnels Filtration Funnels Büchner Funnels
White glazed porcelain.
For filter paper or membrane filtration discs.
Certifications: DIN 12 905 (511-2502, 511-2501 and 511-2500 not according to DIN).