Cleanroom gloves, latex, AK
Supplier: Honeywell Safety
AK1815O/10 AK1815O/11 AK1815O/7 AK1815O/8 AK1815O/9
NORHAK1815O/10EA 156 EUR
NORHAK1815O/10 NORHAK1815O/11 NORHAK1815O/7 NORHAK1815O/8 NORHAK1815O/9
Cleanroom gloves, latex, AK
Gloves
Hand-specific gloves with rolled cuffs and diamond embossed finish. Suitable for cleanroom use and protection from acids and caustics.
  • Non sterile, powder-free, textured finish
  • Outstanding puncture and tear resistance
  • Reduced ionic and extractable contamination
  • Positive grip in wet/dry applications
  • Size embossed instead of inked to eliminate ink contamination
Certifications: EN 388:2016 (1110X), EN ISO 374:2016 Type A (AKLMNOT), EN ISO 374-5:2016 (MICRO-ORGANISMS); Cat. III
Meet IES RP-005.2 (modified) particulate and extractable test. Meet ASTM 739More Product Information