Surgical gloves, latex, sterile, Sempermed® Supreme
Supplier: SEMPERIT TECHN PROD
822751721 822751821 822751801 822751701 822751621
112-0109EA 105 EUR
112-0109 112-0110 112-3134 112-3133 112-0108
Surgical gloves, latex, sterile, Sempermed® Supreme
Gloves
White, pH-neutral surgical gloves with rolled cuff.
  • Powder-free, micro-textured finish
  • The synthetic inner coating provides good skin tolerability and ease of donning without powder even when hands are wet with alcohol
  • High level of comfort and no hand fatigue caused by close-fitting surgical gloves
  • Microrough surface provides effective grip and excellent tactile sensitivity
Length: 270 - 285 mm
Thickness: 0,19 - 0,24 mm (in hand area, measured singly)
Colour: White
EN 455/3, EN 1041, EN 980; AQL 1.0
Certifications: EN 455/3, EN 1041, EN 980; AQL 1.0
Packaging: Left and right glove with cuff turned up in a fibre-free inner package, sealed in a medical peel pack, ozone-tight.