Glove bags
Supplier: abcr
AB104446.0001 AB101721 AB104448.0001 AB104446.0006 AB104448.0006
135-6500EA 0 EUR
135-6500 135-6501 135-6502 ABCRAB104446.0006 ABCRAB104448.0006
Glove bags
Glove Boxes and Glove Bags
These durable PE glove bags effectively eliminate oxygen and moisture during equipment manipulation operations.
  • To be used like a glove box; gloves are integral
  • Equipment is placed in the bag through an entry sleeve, the bag is sealed, connected to the gas port and inflated
  • Economical, since reusable