Chemical resistant gloves, Ultranitril 492
Supplier: MAPA
34492200 34492811 34492816 34492207 34492818 34492209
112-6111EA 46.4 EUR
112-6111 112-6112 112-6113 112-6114 112-6115 112-6116
Chemical resistant gloves, Ultranitril 492
Gloves
These anatomically designed gloves with quality flock lining provide good chemical resistance to hydrocarbon derivatives, alcohols, aromatic and chlorinated solvents.
  • Longer working life: Excellent mechanical resistance (abrasion, puncture)
  • Double layer process: Enhanced chemical protection
  • Recommended for individuals sensitised to natural rubber proteins
Ideal for use in manufacturing industries using cutting oils, metal treatment using solvents, chemical treatments, automotive industries and in the agriculture sector.
Certifications: EN 388 (3101X), EN ISO 374-1 Type A (AJKOPT), EN ISO 374-5 VIRUS, ISO 18889 G2, Cat. III