Chemical resistant gloves, AlphaTec® 08-352 / 08-354
Supplier: Ansell
08-352/10 08-352/8 08-352/9 08-354/10 08-354/8 08-354/9
112-0266EA 17.1 EUR
112-0266 112-3747 112-0267 112-2156 112-2157 112-2158
Chemical resistant gloves, AlphaTec® 08-352 / 08-354
Gloves
Neoprene gloves with exceptional flexibility, offer increased protection against a wide range of chemicals. 100% liquid proof, gloves feature a soft stretchy knit liner with no seams in the working area and added forearm protection.
  • Superb mobility and fingertip sensitivity
  • Excellent grip on wet and slippery materials
  • Extremely comfortable to wear; cushions the hand, absorbs perspiration and prevents chafing
  • Safer handling and reduced hand fatigue mean increased productivity
Typical applications include: Equipment repair & maintenance; filling, blending and charging of raw materials; transferring liquids and solids; loading and unloading of process equipment; cleaning furnaces, distillation pumps, valves or lines and crackers BTX process.
Colour: Green
EN 374 (AKL), EN 388 (3121), Cat. III; AQL 1,0
Certifications: EN ISO 374-1 Type A (AKLMNPT), EN ISO 374-5, EN 388:2016 (3121B), EN 407 (X1XXXX), Cat. III