Industrial gloves, PX130 & PX140
Supplier: Marigold (Ansell)
M10249 M10250 M10251 M10252 M10253 M10258
112-2472EA 31.9 EUR
112-2472 112-2473 112-2474 112-2475 112-2476 112-2481
Industrial gloves, PX130 & PX140
Gloves
Seamless nylon gloves in a lightweight ergonomic design for highly dextrous tasks.
  • Flat dipped polyurethane coating offers unrivalled grip in a variety of conditions
  • Flexible fabric aids dexterity
  • Ventilated back ensures that skin can breathe naturally
Certifications: EN 388 (4131); Cat. II