Knitted gloves, HyFlex® 11-600 / 11-601 / 11-605
Supplier: Ansell
11-600/10 11-600/7 11-600/8 11-600/9 11-605/10 11-605/6 11-605/7 11-605/8 11-605/9 11-600/6 11-601/10 11-601/6 11-601/7 11-601/8 11-601/9 11-600/11 11-601/5 11-600/5
112-0195EA 64.7 EUR
112-0195 112-0197 112-0198 112-0199 112-0200 112-0201 112-0202 112-0203 112-0204 112-0196 112-5073 112-5074 112-5075 112-5076 112-5077 112-4246 ANSE11-601/5 ANSE11-600/5
Knitted gloves, HyFlex® 11-600 / 11-601 / 11-605
Gloves
The stretch knitted nylon liner, together with the formulation of the polyurethane coating, offers flexibility and elasticity.
  • Optimal tactility and dexterity: Ultra-thin with superior tear resistance
  • Polyurethane coating gives a good grip in dry conditions
  • Taint and lint-free
These industrial gloves are ideal for applications where precision and/or clean handling are required and for light and delicate applications, such as assembly of small pieces or for use in cleanrooms.
Certifications: 11-600/11-601: EN 388:2016 (3121A), Cat. II
11-605: Cat. II.
Order Now
SPECIFICATIONS
Model 11-605 11-601 11-600
Environmentally Preferable
Ambidextrous / hand-specific Hand-specific Hand-specific Hand-specific
Disposable / reusable Reusable Reusable Reusable
Gauge 15 15 15
EN norm EN 388:2016 (3121A) EN 388:2016 (3121A)