Dry box gloves, natural rubber (latex)
Supplier: PIERCAN
14800H86/10_U1 14800H76/10_U1 10800H96/10_U1 10750H74/10 10750H84/10 14800H74/10 14800H84/10 18800H104/10 18800H94/10 97800H9E46 97800H9E66 18800H96/10 18800H86/10 10750H86/10 10750H76/10 10750H106/10 18800H84/10 14800H96/10 14800H86/10 14800H76/10 10500H84/10 10500H76/10 14500H76/10 10750H96/10 13800H106/10 13800H76/10 13800H86/10 13800H96/10 10750H94/10 10500H10E4 10500H7E4 10500H9E4 10750H10E4 10500H86/10 10500H96/10 10500H106/10 13800H104/10 13800H74/10 14500H74/10 14500H84/10 14500H104/10 14500H86/10 14500H96/10 14800H9E46 18800H10E45
112-5246EA 93.6 EUR
112-5246 112-5245 112-5218 PIEC10750H74/10 PIEC10750H84/10 PIEC14800H74/10 PIEC14800H84/10 PIEC18800H104/10 PIEC18800H94/10 PIEC97800H9E46 PIEC97800H9E66 PIEC18800H96/10 PIEC18800H86/10 PIEC10750H86/10 PIEC10750H76/10 PIEC10750H106/10 PIEC18800H84/10 PIEC14800H96/10 PIEC14800H86/10 PIEC14800H76/10 PIEC10500H84/10 PIEC10500H76/10 PIEC14500H76/10 PIEC10750H96/10 PIEC13800H106/10 PIEC13800H76/10 PIEC13800H86/10 PIEC13800H96/10 PIEC10750H94/10 PIEC10500H10E4 PIEC10500H7E4 PIEC10500H9E4 PIEC10750H10E4 PIEC10500H86/10 PIEC10500H96/10 PIEC10500H106/10 PIEC13800H104/10 PIEC13800H74/10 PIEC14500H74/10 PIEC14500H84/10 PIEC14500H104/10 PIEC14500H86/10 PIEC14500H96/10 PIEC14800H9E46 PIEC18800H10E45
Dry box gloves, natural rubber (latex)
Gloves
These ambidextrous gauntlets are manufactured from natural rubber that provides good mechanical properties such as tensile strength, perforation and abrasion resistance.
  • Good flexibility and low modulus
  • Low static charge
  • Very high resistance to cold conditions (down to −40 °C)
  • Combination of flexibility and mechanical strength offers protection together with high dexterity for the user
0,4 mm thickness: EN 388 (X100), EN 374-1, EN 421; Cat. III
0,6 mm thickness: EN 388 (X111), EN 374-1, EN 421; Cat. III
Certifications: 0,4 mm thickness: EN ISO 374-1 Type C, EN ISO 374-5 VIRUS, EN 421; Cat. III
0,6 mm thickness: EN 388:2016 (XX11X), EN ISO 374-1 Type C, EN ISO 374-5 VIRUS, EN 421; Cat. III