General purpose gloves, Perfect Poly® black
Supplier: HONEYWELL SAFETY
2400251/6 2400251/8 2400251/9 2400251/10 2400251/7
112-2275EA 6.4 EUR
112-2275 112-2277 112-2278 112-2279 112-2276
General purpose gloves, Perfect Poly® black
Gloves
Knitted gloves made of lightweight polyamide.
  • Grey polyurethane coating on palm and fingertips
  • Excellent dexterity
  • High abrasion resistance
Suitable for fine handlings in dry and heavily soiled environments.
Certifications: EN 388:2016 (4121X); Cat. II.