Tissue grinders, conical
Supplier: DWK Life Sciences
358133 358137 358141 358145 358132 358144 358143 358140 358139 358136 358135 358131
432-1290EA 162 EUR
432-1290 432-1291 432-1292 432-1293 432-1294 432-5062 432-5061 432-5060 432-5059 432-5058 432-5057 432-5056
Tissue grinders, conical
Homogenisers Tissue Grinders
Mortar in borosilicate glass, PTFE pestle with stainless steel rod. Handheld homogeniser with conical surface, which allows initial size reduction, followed by passage through the cylindrical section for the final homogenisation step. Grinding efficiency is improved and less time is required as compared to the Tenbroeck and Potter-Elvehjem designs. Recommeded for soft tissues such as brain.
  • Clearance between pestle and tube is 0,1 to 0,15 mm
The stainless steel rod Ø 6,3 mm is long enough to fit into the chuck of overhead stirrers.
Certifications: More Product Information