Disposable particle filtering respirators, FFP1/FFP2/FFP3, 4000 series
Supplier: HONEYWELL SAFETY
1005605 1005608 1005611 1005614 1005630
111-1273EA 39.5 EUR
111-1273 111-1274 111-1277 111-1278 111-1279
Disposable particle filtering respirators, FFP1/FFP2/FFP3, 4000 series
Respirators Disposable Respirators Particulate Respirators
Individually wrapped fold-flat respirators with the exclusive features of the Willson 5000 series: ultra-soft seal, new exhalation valve, latex, PVC, silicone-free materials.
  • Five models in a fold-flat style presented in hygienic individual sachets printed with user instructions
  • Can be conveniently stored in a pocket
  • Fold and hold mechanism prevents contaminants from entering the masks when not in use
  • Chin piece specifically designed to allow users normal movement during speech
EN149:2001
Certifications: EN 149:2001