Particulate disc filters 2000 series for masks, 6000/7000/7500 series
Supplier: 3M
2125 2128 2135 2138 2128UK
111-0069EA 161 EUR
111-0069 111-0070 111-0055 111-0056 111-2181
Particulate disc filters 2000 series for masks, 6000/7000/7500 series
Respirators Reusable Respirators Respirator Filters
These mask filters are light, comfortable and provide reliable protection from particulates. When using with 3M™ 6000 series masks, a 3M™ 502 adapter is required.
  • Low breathing resistance
  • Allow a very wide field of vision
  • Bayonet fitting system ensures precise and secure locking
EN 143
Certifications: EN 143