Measuring cylinders, tall form, class A, borosilicate glass
Supplier: HIRSCHMANN
2240194 2240154 2240160 2240170 2240175 2240180 2240186 2240190 2240193
612-3072EA 133 EUR
612-3072 612-1517 612-1518 612-1519 612-1520 612-1521 612-1522 612-1523 612-1524
Measuring cylinders, tall form, class A, borosilicate glass
Cylinders Measuring Cylinders
DURAN®, borosilicate glass, tall form, class A.
  • Calibrated to contain 'ln', certificate of conformity (DE-M marking) with dated batch identification available via website: (www.hirschmannlab.de)
  • Hexagonal glass base, with spout
  • Blue graduations with ring marks at major graduations
DIN EN ISO 4788