Measuring cylinders, tall form, class B, PP, Nalgene®
Supplier: Thermo Fisher Scientific
3662-0010 3662-0025 3662-0050 3662-0100 3662-0250 3662-0500 3662-1000 3662-2000 3662-4000
612-4020EA 18.3 EUR
612-4020 612-4021 612-4022 612-4023 612-4024 612-4025 612-4026 612-4027 612-4028
Measuring cylinders, tall form, class B, PP, Nalgene®
Cylinders Measuring Cylinders
PPCO, translucent, tall form, class B.
  • Large blue hexagonal base aids stability
  • Large spout, blue moulded graduations
  • Can be chemically sterilised without affecting accuracy, autoclaving will affect accuracy
ISO 6706, CFR21
Certifications: ISO 6706, CFR21