Measuring cylinders, tall form, PMP
Supplier: KARTELL
2575 2570 2571 2572 2573 2574 2576 2577
612-4073EA 44.4 EUR
612-4073 612-4068 612-4069 612-4070 612-4071 612-4072 612-4074 612-4075
Measuring cylinders, tall form, PMP
Cylinders Measuring Cylinders
PMP (TPX®), transparent, tall form, autoclavable.
  • Pouring spout, pentagonal base and moulded blue graduations
  • No meniscus, chemically non absorbent, non wetting surface, excellent chemical resistance
  • Can be used with liquids up to +170 °C
ISO 6706, BS 5404 Part 2
Certifications: ISO 6706, BS 5404 Part 2