Measuring cylinders, tall form, PP
Supplier: KARTELL
2566 2560 2561 2562 2563 2564 2565 2567
612-4066EA 41.7 EUR
612-4066 612-4060 612-4061 612-4062 612-4063 612-4064 612-4065 612-4067
Measuring cylinders, tall form, PP
Cylinders Measuring Cylinders
PP, translucent, tall form.
  • Pouring spout, pentagonal base and blue graduations
  • Autoclavable at +121 °C, can withstand temperatures up to +100 °C during continuous work
ISO 6706, BS 5404 Part 2
Certifications: ISO 6706, BS 5404 Part 2