Measuring cylinders, tall form, class B, PP
Supplier: VITLAB
652081 653081 646081 651081 647081 648081 649081 650081
612-4314EA 21.7 EUR
612-4314 612-4316 612-4302 612-4312 612-3787 612-3788 612-3789 612-3790
Measuring cylinders, tall form, class B, PP
Cylinders Measuring Cylinders
PP, translucent, tall form, autoclavable, class B.
  • Calibrated to contain 'ln'
  • Hexagonal base, spout
  • Ring-moulded graduations, overprinted in blue
Certifications: DIN 12681 / ISO 6706