Measuring cylinders, low form, class B, borosilicate glass, SILBERBRAND ETERNA
Supplier: Brand
42062 42064 42038 42048 42054 42028 42008 42020
612-5116EA 54.6 EUR
612-5116 612-5117 612-5099 612-5100 612-5115 612-5098 612-5096 612-5097
Measuring cylinders, low form, class B, borosilicate glass, SILBERBRAND ETERNA
Cylinders Measuring Cylinders
Borosilicate glass 3.3, low form, class B.
  • Calibrated to contain (TC, In)
  • Hexagonal base, spout
  • Marks and inscriptions in ETERNA amber stain, short graduation marks
DIN  EN ISO 4788
Certifications: DIN  EN ISO 4788