Measuring cylinders, tall form, class B, borosilicate glass
Supplier: KAVALIER GLASS
432140819 432141130 432141444 432141546 432141238 432141343
612-3770EA 21 EUR
612-3770 612-3773 612-3776 612-3777 612-3774 612-3775
Measuring cylinders, tall form, class B, borosilicate glass
Cylinders Measuring Cylinders
Borosilicate glass, tall form, class B.
  • Hexagonal base
  • White graduations
Certifications: ISO 4788, DIN 12 680-1