Plates, Nunc-Immuno™ TSP (transferable solid phases)
Supplier: Thermo Fisher Scientific
445497 476635
735-0022EA 424 EUR
735-0022 735-0074
Plates, Nunc-Immuno™ TSP (transferable solid phases)
Microplate and Petri Dish Replicators
PS. For simultaneous testing of analytes using ELISA technique. Also available sterile for screening of hybridoma cultures for antibody production.
  • Uniform adsorption properties
  • Plates have 96 round bottom wells with total surface area of 95 mm²
  • Sterile plates supplied mounted in the tray
MaxiSorp™ surface has a high affinity to molecules with mixed hydrophilic/hydrophobic domains.
Colour: Clear
Colour: Clear