Accessories for Microplates, Nunc™
Supplier: Thermo Fisher Scientific
5500 5550
735-0189EA 259 EUR
735-0189 735-0488
Accessories for Microplates, Nunc™
Microplates Immunology Microplates and Strips
Designed for use with Thermo Scientific™ microplates. Type A lids are suitable for Immulon®, Microfluor® and Microlite™ ranges.
  • Type A lids are suitable for Immulon®, Microfluor® and Microlite™ ranges