Plates, collagen I coated, Nunc™
Supplier: Thermo Fisher Scientific
152034 152036 152038 152040
735-0226EA 726 EUR
735-0226 734-1183 734-1185 734-1187
Plates, collagen I coated, Nunc™
Microplates Cell Culture Plates
PS ready to use plates pre-coated with collagen I, with lid.
  • Consistent growth surface from each lot ensures guaranteed performance
  • High quality surface promotes cell attachment, growth and differentiation in serum-free and serum-containing media
  • Stable at room temperature
W×L: 128×86 mm.
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Environmental certification NAR Certificate of Retirement