Modules, Nunc-Immuno™, BreakApart™
Supplier: Thermo Fisher Scientific
473539 473768
735-0062EA 705 EUR
735-0062 735-0065
Modules, Nunc-Immuno™, BreakApart™
Microplates Immunology Microplates and Strips
PS. Designed for solid phase immunoassays, for example, ELISA techniques. Available with MaxiSorp™ or PolySorp™ surface chemistries, the product consists of a frame with 12 carriers, each holding a breakable strip of 8 wells. All MaxiSorp™ modules are certified for binding homogeneity. Framed modules with alphanumeric orientation (external dimensions: 128×86 mm) fit all standard equipment.
  • C-shaped well bottom (flat with curved edges) for optimal washing and liquid retrieval
  • Push-out tool supplied with every case of 60 modules
  • Colour coded modules facilitate assay identification (please enquire for availability)
  • Free-standing modules
Recommended working volume: 250 µl.
Recommended working volume: 250 µl