PCR plates, 96-well, non-skirted, Thermo-Fast®
Supplier: Thermo Fisher Scientific
AB-0600 AB-0600/W AB-0600/B AB-0600/G AB-0600/P AB-0600/R AB-0600/Y AB-0600/L
732-4828EA 153 EUR
732-4828 732-5008 732-5003 732-5004 732-5006 732-5007 732-4829 731-0276
PCR plates, 96-well, non-skirted, Thermo-Fast®
Microplates PCR Plates
PP. Suitable for all 0,2 ml thermal cycler blocks and ABI PRISM® 310, 3100 and 3700 capillary sequencers. Available in opaque white for luminescent based procedures.
  • Maximum well volume 0,3 ml when used with adhesive and heat seals
  • Cut corner (H1)
  • Can be sealed using Thermo-Mats™, adhesive films and foils, heat seals and flat or domed cap strips
  • Certified free from DNase, RNase and human genomic DNA