Culture slides, Falcon®
Supplier: Corning
354104 354108 354114 354118
734-0401EA 678.2 EUR
734-0401 734-0402 734-0088 734-0089
Culture slides, Falcon®
Slides Chamber Slides
Soda-lime glass slide, PS vessel, lid and tool for loosening chamber.
  • Chamber easily and safely removed using the disposable safety removal tool
  • Pressure-sensitive, biocompatible, acrylic-adhesive gasket remains with the vessel after removal, not on the slide, facilitating further processing or placement of coverslips
  • Blue hydrophobic border defines cell culture areas
  • Wells numbered for easy identification
  • Trays designed for incubator use
Falcon® CultureSlides allow cells to be cultured and then analysed on a glass microscope slide. Cells are grown in a plastic chamber attached to a specially prepared microscope slide. Cells can be fixed and stained in situ without disruption of the cell monolayer.
W×L: 25×75 mm with 1,2 mm bevelled edge.
Ordering information: Supplied with removal tool and key.