Mop heads, cleanroom, PurSPONGE
Supplier: VWR Collection
129-0048EA 845 EUR
129-0048 129-0051 129-0052 129-0053 129-0054 129-2504
Mop heads, cleanroom, PurSPONGE
Mops
Mops suitable for wet wiping in sterile areas and cleanrooms. One-piece, made from highly absorbent polyurethane. These coated mop heads are ideal for disinfecting rough or sharp edged surfaces, while still ensuring excellent take-up of liquids.
  • Models available for use with Perfex or Roll-O-Matic® handles
  • Perfex model made from PU (on PE base), coated or uncoated. Roll-O-Matic® model made from highly absorbent PU sponge with clamp, available with or without coating
Models available for use with Perfex or Roll-O-Matic® handles:
-Perfex model, made from PU (on PE base), coated or uncoated
-Roll-O-Matic® model made from highly absorbent PU sponge with clamp, available with or without coating
Ordering information: Perfex or Roll-O-Matic® mop handles are not included. These must be ordered separately.