VWR®, Rigid Sleeves for Standard Ground Joints
Supplier: VWR Collection
201-0039EA 83.5 EUR
201-0039 201-0040 201-0041 201-0042 201-0043 201-0044 201-0045 201-0046 201-0048
VWR®, Rigid Sleeves for Standard Ground Joints
NS Joint Sleeves and Connectors Joint Sleeves
PTFE, with grip.
  • Very good chemical resistance
  • Gas, liquid and vacuum-tight
  • Temperature resistant from −200 to +280 °C
For fitting onto standard ground cones. Sturdy construction with protruding grip so sleeves can be detached easily. With sealing rings on the outside of the ground socket, which prevent ground joints from seizing and give a very good seal. Grease-free connection; no grease residue to contaminate sample.