Aspirating pipettes, Falcon®
Supplier: Corning
357558 357501
734-0456EA 58.05 EUR
734-0456 612-2458
Aspirating pipettes, Falcon®
Pipettes Aspirating Pipettes
PS pipettes, non plugged, non graduated, disposable, sterile.
  • Safer alternative to glass Pasteur pipettes
  • Non pyrogenic
  • Individually packed to prevent contamination
Packaging: Individually packed in a thermo-formed paper/plastic wrap.