Transfer pipettes, Falcon®
Supplier: Corning
357524 357575
734-0342EA 94.52 EUR
734-0342 734-0354
Transfer pipettes, Falcon®
Pipettes Pasteur/Transfer Pipettes
PE, disposable.
  • 1 and 2 ml graduations
  • Small tip ensures consistent reproduction of drop size
Ordering information: Nonsterile pipettes are bulk packaged. Sterile pipettes are individually wrapped.