Pipette controller, pipetus®-standard
Supplier: HIRSCHMANN
9903000 9904000 9905000 9906000 9904010 9903010
612-2602EA 286 EUR
612-2602 612-2603 HIRS9905000 HIRS9906000 HIRS9904010 HIRS9903010
Pipette controller, pipetus®-standard
Pipettes Pipette Controllers
Pipette controller for pipettes from 0,1 to 100 ml.
  • Optimum safety and excellent convenience of use
  • Pipettor is lighter and easy to use because the pump is separated from the handle
  • Autoclavable, detachable handle for the pipette holder is equipped with two buttons for filling and emptying
This electric pipettor is equipped with a 0,2 µm integrated water-repellent filter (for better durability) and a safety valve to protect the equipment from the introduction of hazardous (contaminated or radioactive) liquids.
Ordering information: The completeMore Product Information