VWR®, Pipette Tips, Standard
Supplier: VWR Collection
613-1050EA 11.8 EUR
613-1050 613-1051 613-1053 613-1054 613-1055 613-1056 613-1057 613-3019
VWR®, Pipette Tips, Standard
Pipette Tips
These universal tips are compatible with most micropipettes.
  • Non sterile, autoclavable
  • Smooth inside surface finish for minimum retention of dispensed liquid
Colour: Transparent (0,5 - 10 µl), yellow (2 - 200 µl and 5 - 200 µl), blue (200 - 1000 µl)
Ordering information: Supplied loose or in racks holding 96 tips.