Robotic tips for Tecan®
Supplier: Thermo Fisher Scientific
901-262 904-251 901-252 903-262 903-261 903-021 904-262 904-261 904-021 901-261 902-262 902-261 902-021 902-253 903-011G 903-251G 903-252G 904-011G 904-251G 904-252G 903-252 903-251 903-011 904-252 904-011 901-251 901-011 902-252 902-251 902-011 901-253 903-253 904-253 901-021 908-261 907-261 908-021 908-262 907-262 902-262XP 906-262 906-261 902-261XP 906-021 907-021 903-033
736-0045EA 188 EUR
736-0045 736-0061 736-0043 736-0058 736-0010 736-0009 736-0066 736-0012 736-0011 736-0006 736-0050 736-0008 736-0007 736-0049 736-0052 736-0054 736-0056 736-0060 736-0062 736-0064 736-0055 736-0053 736-0051 736-0063 736-0059 736-0042 736-0041 736-0048 736-0047 736-0046 736-0044 736-0057 736-0065 736-0005 732-1527 732-1528 732-1529 732-1530 732-1531 MLBP902-262XP 732-1532 732-1533 MLBP902-261XP 732-1534 732-1535 MLBP903-033
Robotic tips for Tecan®
Pipette Tips
High quality tips, especially intended for research in molecular biology laboratories.
  • Transparent or black PP
  • Tips with ART® filters: Eliminate any risk of cross-contamination between samples; designed for applications with DNA, RNA as well as radioactive and infectious samples
  • Certified free of RNase, DNase, DNA and pyrogens
  • Sterile tips: Sterilised by gamma radiation
  • Wide aperture tips: Designed for applications with more viscous samples
Ordering information: Available in boxes of 10 racks or 50 racks (Econopak).