Glass density bottles, calibrated, BLAUBRAND®
Supplier: Brand
43305 43308 43320 43328 43338
614-0701EA 48 EUR
614-0701 614-0702 614-0703 614-0704 614-0705
Glass density bottles, calibrated, BLAUBRAND®
Specific Gravity Bottles
Borosilicate glass 3,3, DIN ISO 3507, Gay-Lussac type.
  • Bottle and stopper (with capillary) are marked with unique matching identification number
  • Individual certificate included
  • Reference temperature: 20 °C
The density bottles (pycnometers) are individually adjusted. The nominal capacity is indelibly engraved on each bottle. Each bottle is calibrated with its own stopper. The volume in cm3 is specified to a precision of 3 decimal places.