Racks, microcentrifuge tube
Supplier: KARTELL
563-03 563-04 563-10 563-06
732-2000EA 28.3 EUR
732-2000 732-2001 732-2003 732-2002
Racks, microcentrifuge tube
Racks Tube Racks
PP, autoclavable.
  • Versatile racks, can be used dry, in water baths or in freezers
  • Stackable when empty or full
  • Alphanumeric grid reference for easy sample identification
Can accommodate 100×1,5 ml microtubes with closed caps or without caps, or 50×1,5 ml microtubes with caps open.