Microtube racks, floating, Nalgene®
Supplier: Thermo Fisher Scientific
5974-0404 5974-1015 5974-4015
211-7312EA 50.7 EUR
211-7312 211-7314 211-7313
Microtube racks, floating, Nalgene®
Racks Tube Racks
PP, floating.
  • Temperature resistant from −70 to +100 °C, autoclavable
  • Alphanumeric labelling of rack positions for easy identification of samples
  • Handles enable the racks to be carried, shifted and removed easily
Ideal for thawing, cooling or incubating samples at low or high temperatures. Racks float even when full. Tubes are kept below the rim. Round racks 211-7313 and 213-7314 fit in 400 and 1000 ml low-form beakers.