Test tube racks, Unwire™, Nalgene®
Supplier: Thermo Fisher Scientific
5970-0013 5970-0016 5970-0020 5970-0025 5970-0030 5970-0113 5970-0116 5970-0120 5970-0125 5970-0130 5970-0213 5970-0216 5970-0220 5970-0225 5970-0230 5970-0313 5970-0316 5970-0320 5970-0325 5970-0330 5970-0413 5970-0416 5970-0420 5970-0425 5970-0430 5970-0513 5970-0516 5970-0520 5970-0525 5970-0530
212-8111EA 33.4 EUR
212-8111 212-8121 212-8131 212-8141 212-8151 212-8100 212-8101 212-8132 212-8102 212-8103 212-8113 212-8123 212-8133 212-8143 212-8153 212-8114 212-8124 212-8134 212-8144 212-8154 212-8115 212-8125 212-8135 212-8145 212-8155 212-8116 212-8126 212-8136 212-8146 212-8156
Test tube racks, Unwire™, Nalgene®
Racks Tube Racks
ResMer™, colour coded.
  • Heat resistant up to +135 °C, autoclavable
  • Good chemical resistance
The ResMer™ manufacturing technology improves the shelf life of these test tube racks under the harshest conditions such as autoclaving, cleaning in dishwashers, chemical disinfection and use in freezing conditions. They are unmistakable with durable and brighter colours. The racks do not float, so they are also very suitable for use in ice or water baths.