Reaction vessels, cylindrical, with flat flange, DURAN®
Supplier: DWK Life Sciences
243902408 243903601 243904409 243905405 243906307 243907106 243907603 243908608
273-0111EA 48.9 EUR
273-0111 273-0112 201-0113 201-0114 201-0115 201-0116 201-0117 201-0118
Reaction vessels, cylindrical, with flat flange, DURAN®
Reaction Vessels Reaction Vessels, Cylindrical Shape
DURAN®, borosilicate glass 3.3, clear, with round bottom.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With flat flange (DN) with groove for vacuum seal. Maximum operating pressure at +250 °C: 2,5 bar for 100 and 250 ml; 1,5 bar for 500, 1000 and 2000 ml; 1,0 bar for 4000 and 6000 ml; and 0,5 bar for 10000 ml.
*Vessel cylindrical throughout and has a flat bottom.
Caution: Important: At a maximum operating temperature of +250 °C and at permissibleMore Product Information