Flat flange beaker, DURAN®
Supplier: DWK Life Sciences
243915406 243916805 243945409 243946302 243946808 243916308
201-0146EA 143 EUR
201-0146 273-0148 201-0141 201-0142 201-0143 273-0147
Flat flange beaker, DURAN®
Reaction Vessels Reaction Vessels, Beaker Shape
DURAN® borosilicate glass 3.3, clear, with laboratory flat flange (DN) with groove for vacuum use. Maximum operating pressure at 250 °C: 0,5 bar.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Caution: Important: At a maximum operating temperature of 250 °C and at permissible operating pressure, the temperature difference in the glass wall of the flat flange reaction vessels may not exceed 30 °C.