Stand bases
Supplier: USBECK Laborgeräte
2012 2013 2014-2144 342012 342014 342015 2015
241-0366EA 70 EUR
241-0366 241-7175 USBE2014-2144 241-7101 241-7102 241-0348 USBE2015
Stand bases
Retort Stands
Stainless steel. Firm, non slip stand for a wide variety of uses. With an M10 threaded hole on face side and adjustable feet for uneven surfaces.
  • Rubber feet for a firm footing
  • Smooth surfaces for easy, fast cleaning
  • Robust
DIN 12892/55
Certifications: DIN 12892/55