Minor skin graft blade, No. SG3
Supplier: Swann-Morton
233-0053EA 43.9 EUR
233-0053
Minor skin graft blade, No. SG3
Scalpels and Scalpel Assemblies Scalpel Blades
Stainless steel. Originally developed for use with skin grafts, these blades are also suitable for a wide range of other applications.
  • Fits standard No. 3 surgical handle (233-5454)
  • Sterile
  • Individually packed
Originally developed for use with skin grafts, these blades are also suitable for a wide range of other applications.
  • With blade 50 mm in length
  • For scalpel handles No. 3 (233-5454)
  • Individually sterile-packed
Ideal for use on the hands and feet of small children given its practical size advantage compared to a standard skin grafting knife. Also suitable for use within Histology departments for the fine sectioning of the pathology blocks.
Packaging: Packed in individual foil packs.