Scalpels, ceramic
Supplier: SLG Kunststoff
393916 394173 394235 394035
233-5091EA 163 EUR
233-5091 233-5092 233-5093 233-5094
Scalpels, ceramic
Scalpels and Scalpel Assemblies Scalpels
Particularly high quality and durable hard ceramic material for long lasting, excellent sharpness. No metal parts, thus also suitable for trace analysis. Super-lightweight, sturdy, ergonomic handle for safe and easy handling.
  • Anti-magnetic, anti-static, corrosion-free, dishwasher safe, chemically resistant to aggressive media
  • Extremely heat resistant, low thermal conductivity
  • Regrinding is not necessary