Thread and vessel scissors
Supplier: HAMMACHER KARL
HWB010-11 HWB040-13 HWB042-14 HWB044-16
233-1117EA 102 EUR
233-1117 233-0083 233-1106 233-0084
Thread and vessel scissors
Scissors
Stainless steel 18/10, anti-magnetic.
  • Sharp-blunt
  • Ideal for a wide range of laboratory applications
  • Acid-resistant
  • Autoclavable