Seals for automated microplate heat sealer, ALPS™ 3000
Supplier: THERMO MATRIX TECHNOLOGIES
AB-3559 AB-3720 AB-3738 AB-3739 AB-3797 AB-3799
732-0842EA 1835 EUR
732-0842 732-0843 732-0844 732-0845 732-0846 732-0847
Seals for automated microplate heat sealer, ALPS™ 3000
Seals Microplate Seals
Thermo-Seal
Foil laminate; very strong seal that can only be peeled using foil stripper; can be pierced with a single pipette tip or Pierce Plate; good solvent resistance; recommended for use with DMSO
Compatible plate material: PP
Sealing conditions: 1,5 to 2,5 seconds at 165 to 170 °C
Seal integrity range: −80 to +120 °C
Applications: long term storage including storage at low temperatures; transportation and high temperature applications such as PCR, including use in waterbath thermal cyclers
Easy Pierce
Foil material; easily pierced; peelable bond that can be removed by hand; second seal can be applied over existing seal; recommended for use with DMSO
Compatible plate material: PP, PE
Sealing conditions: 0,5 to 3,0 seconds at 165 to 175 °C
Seal integrity range: −80 to +80 °C and up to 120 °C with cycler lid pressure
Applications: well suited for 384-well formats where piercing is a useful method for sample retrieval; second seal can be applied over existing seal; suitable for PCR with screw-down lid thermal cyclers
Easy Pierce 20 µm
Thin foil material; easily pierced; peelable bond that can be removed by hand; good solvent resistance; recommended for use with DMSO
Compatible plate material: PP, PE
Sealing conditions: 0,5 to 2,0 seconds at 165 to 175 °C
Seal integrity range: −80 to +80 °C and up to 120 °C with cycler lid pressure
Applications: well suited for 384-well formats where piercing is a useful method for sample retrieval; suitable for PCR with screw-down lid thermal cyclers
Clear Seal
Thin polymer material; able to be pierced; peelable seal when layered; good optical clarity; some solvent resistance
Compatible plate material: PP, PS, PE
Sealing conditions: 1,5 seconds at 170 °C
Seal integrity range: −80 to +80 °C and up to 120 °C with cycler lid pressure
Applications: suitable for fluorescence and colorimetric applications; suitable for PCR applications with screw-down or clip-down heated lid thermal cyclers only
Clear Seal Diamond
Thin polymer material; peelable seal; good optical clarity; some solvent resistance
Compatible plate material: PP, PS, PE, COC
Sealing conditions: 1,5 to 3,0 seconds at 170 °C
Seal integrity range: −80 to +120 °C
Applications: ideal for fluorescence and colorimetric applications; suitable for PCR applications with screw-down or clip-down heated lid thermal cyclers only